Learn More
Trajan Scientific Column BPX5 QC3 5 in 1/EA
SDP
Supplier:  Trajan Scientific 054153CCS
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 30 m length BPX5 GC capillary column on 5 cage
Catalog No. 50-004-94688