Learn More
Trajan Scientific Column BPX5 QC3 Medical 1/EA
SDP
Catalog No. 5000494332
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 30 m length BPX5 GC capillary column
Catalog No. 50-004-94332 Supplier Trajan Scientific Supplier No. 054153
0 32 mm ID x 1 0 m film thickness x 30 m length BPX5 GC capillary column