Learn More
Trajan Scientific Column BPX5 QC3 Medical 1/EA
SDP
Catalog No. 5000494302
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 12 m length BPX5 GC capillary column
Catalog No. 50-004-94302 Supplier Trajan Scientific Supplier No. 054127
0 32 mm ID x 1 0 m film thickness x 12 m length BPX5 GC capillary column