Learn More
Trajan Scientific Column BPX5 QC3 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054129
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 50 m length BPX5 GC capillary column
Catalog No. 50-004-94254