Learn More
Trajan Scientific Column BPX5 QC3 Medical 1/EA
SDP
Catalog No. 5000494643
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 25 m length BPX5 GC capillary column
Catalog No. 50-004-94643 Supplier Trajan Scientific Supplier No. 054119
0 32 mm ID x 0 25 m film thickness x 25 m length BPX5 GC capillary column