Learn More
Trajan Scientific Column BPX5 QC3 Medical 1/EA
SDP
Catalog No. 5000494617
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 50 m length BPX5 GC capillary column
Catalog No. 50-004-94617 Supplier Trajan Scientific Supplier No. 054126
0 32 mm ID x 0 5 m film thickness x 50 m length BPX5 GC capillary column