Learn More
Trajan Scientific Column BPX5 QC5 Medical 1/EA
SDP
Catalog No. 5000493635
Encompass_Preferred
0 53 mm ID x 0 5 m film thickness x 30 m length BPX5 GC capillary column
Catalog No. 50-004-93635 Supplier Trajan Scientific Supplier No. 0541345
0 53 mm ID x 0 5 m film thickness x 30 m length BPX5 GC capillary column