Learn More
Trajan Scientific Column BPX5 QC5 Medical 1/EA
SDP
Catalog No. 5000493652
Encompass_Preferred
0 53 mm ID x 1 0 m film thickness x 25 m length BPX5 GC capillary column
Catalog No. 50-004-93652 Supplier Trajan Scientific Supplier No. 054131
0 53 mm ID x 1 0 m film thickness x 25 m length BPX5 GC capillary column