Learn More
Trajan Scientific Column BPX50 QC1.5 Medical 1/EA
SDP
Catalog No. 5000494285
Encompass_Preferred
0 15 mm ID x 0 15 m film thickness x 30 m length BPX50 GC capillary column
Catalog No. 50-004-94285 Supplier Trajan Scientific Supplier No. 054741
0 15 mm ID x 0 15 m film thickness x 30 m length BPX50 GC capillary column