Learn More
Trajan Scientific Column BPX50 QC2.5 Medical 1/EA
SDP
Catalog No. 5000494458
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 60 m length BPX50 GC capillary column
Catalog No. 50-004-94458 Supplier Trajan Scientific Supplier No. 054752
0 25 mm ID x 0 25 m film thickness x 60 m length BPX50 GC capillary column