Learn More
Trajan Scientific Column BPX70 QC1 Medical 1/EA
SDP
Catalog No. 5000494182
Encompass_Preferred
0 1 mm ID x 0 2 m film thickness x 10 m length BPX70 GC capillary column
Catalog No. 50-004-94182 Supplier Trajan Scientific Supplier No. 054600
0 1 mm ID x 0 2 m film thickness x 10 m length BPX70 GC capillary column