Learn More
Trajan Scientific Column BPX70 QC2.5 5 in 1/EA
SDP
Catalog No. 5000493765
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length BPX70 GC capillary column on 5 cage
Catalog No. 50-004-93765 Supplier Trajan Scientific Supplier No. 054622CCS
0 25 mm ID x 0 25 m film thickness x 30 m length BPX70 GC capillary column on 5 cage