Learn More
Trajan Scientific Column BPX70 QC2.5 Medical 1/EA
SDP
Catalog No. 5000494105
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 60 m length BPX70 GC capillary column
Catalog No. 50-004-94105 Supplier Trajan Scientific Supplier No. 054623
0 25 mm ID x 0 25 m film thickness x 60 m length BPX70 GC capillary column