Learn More
Trajan Scientific Column BPX70 QC2 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054603
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 50 m length BPX70 GC capillary column
Catalog No. 50-004-94647