Learn More
Trajan Scientific Column BPX70 QC2 Medical 1/EA
SDP
Catalog No. 5000494600
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 30 m length BPX70 GC capillary column
Catalog No. 50-004-94600 Supplier Trajan Scientific Supplier No. 054612
0 22 mm ID x 0 25 m film thickness x 30 m length BPX70 GC capillary column