Learn More
Trajan Scientific Column BPX70 QC3 5 in 1/EA
SDP
Supplier:  Trajan Scientific 054616CCS
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 30 m length BPX70 GC capillary column on 5 cage
Catalog No. 50-004-93915