Learn More
Trajan Scientific Column BPX70 QC3 5 in 1/EA
SDP
Catalog No. 5000494252
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 60 m length BPX70 GC capillary column on 5 cage
Catalog No. 50-004-94252 Supplier Trajan Scientific Supplier No. 054617CCS
0 32 mm ID x 0 25 m film thickness x 60 m length BPX70 GC capillary column on 5 cage