Learn More
Trajan Scientific Column BPX70 QC3 Medical 1/EA
SDP
Catalog No. 5000494187
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 25 m length BPX70 GC capillary column
Catalog No. 50-004-94187 Supplier Trajan Scientific Supplier No. 054606
0 32 mm ID x 0 25 m film thickness x 25 m length BPX70 GC capillary column