Learn More
Trajan Scientific Column BPX70 QC3 Medical 1/EA
SDP
Catalog No. 5000493837
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 30 m length BPX70 GC capillary column
Catalog No. 50-004-93837 Supplier Trajan Scientific Supplier No. 054616
0 32 mm ID x 0 25 m film thickness x 30 m length BPX70 GC capillary column