Learn More
Trajan Scientific Column BPX70 QC5 Medical 1/EA
SDP
Catalog No. 5000494703
Encompass_Preferred
0 53 mm ID x 0 5 m film thickness x 30 m length BPX70 GC capillary column
Catalog No. 50-004-94703 Supplier Trajan Scientific Supplier No. 054620
0 53 mm ID x 0 5 m film thickness x 30 m length BPX70 GC capillary column