Learn More
Thermo Scientific COLUMN,CG19-4ML ION PK,2X500ML
Catalog No. NC1164041 Shop All Thermo Scientific Products
Encompass
Highest resolution, through the use of smaller resin particles, of common cations and small polar amines in complex sample matrices-Higher peak efficiency and increased peak resolution for more consistent integration and more reliable results-Easily transfer applications from the Dionex IonPac CS19 column to the 4 µm column due to the similar selectivity
Catalog No. NC1164041 Supplier Thermo Scientific Supplier No. 078839
Highest resolution, through the use of smaller resin particles, of common cations and small polar amines in complex sample matrices-Higher peak efficiency and increased peak resolution for more consistent integration and more reliable results-Easily transfer applications from the Dionex IonPac CS19 column to the 4 µm column due to the similar selectivity