Learn More
Chemglass Life Sciences Column, Chromatography, 50/30 Socket Joint, 2 1/2in ID X 18in E.L., 4mm Stpk
SDP
Catalog No. 501531051
Encompass_Preferred
Column, Chromatography, 50/30 Socket Joint, 2 1/2in ID X 18in E.L., 4mm Stopcock. Similar to CG-1198 but with a COARSE porosity disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with optional CG-1201 flow controller and CG-1200 reservoir. Lower end of the column has a round bottom. Connections between the column, reservoir and flow control adapter are spherical joints connected with CG-150 Stainless Steel pinchclamps.
Catalog No. 50-153-1051 Supplier Chemglass Life Sciences Supplier No. CG119935
Add to Cart
Add to Cart
Column, Chromatography, 50/30 Socket Joint, 2 1/2in ID X 18in E.L., 4mm Stopcock. Similar to CG-1198 but with a COARSE porosity disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with optional CG-1201 flow controller and CG-1200 reservoir. Lower end of the column has a round bottom. Connections between the column, reservoir and flow control adapter are spherical joints connected with CG-150 Stainless Steel pinchclamps.