Learn More
Chemglass Life Sciences Column, Chromatography, 24/40 Outer Rodaviss Joint, 1 1/2in ID X 8in E.L., 2mm Stpk
SDP
Catalog No. 501531043
Encompass_Preferred
Column, Chromatography, 24/40 Outer Joint, 1/2in ID X 18in E.L., 2mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.
Catalog No. 50-153-1043 Supplier Chemglass Life Sciences Supplier No. CG119213
Column, Chromatography, 24/40 Outer Joint, 1/2in ID X 18in E.L., 2mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.