Learn More
Chemglass Life Sciences Column, Chromatography, 35/20 Socket Joint, 3/4in ID X 18in E.L., 2mm Stpk
SDP
Catalog No. 501531048
Encompass_Preferred
Column, Chromatography, 35/20 Socket Joint, 3/4in ID X 18in E.L., 2mm Stopcock. Similar to CG-1198 but with a COARSE porosity disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with optional CG-1201 flow controller and CG-1200 reservoir. Lower end of the column has a round bottom. Connections between the column, reservoir and flow control adapter are spherical joints connected with CG-150 Stainless Steel pinchclamps.
Catalog No. 50-153-1048 Supplier Chemglass Life Sciences Supplier No. CG119917
Column, Chromatography, 35/20 Socket Joint, 3/4in ID X 18in E.L., 2mm Stopcock. Similar to CG-1198 but with a COARSE porosity disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with optional CG-1201 flow controller and CG-1200 reservoir. Lower end of the column has a round bottom. Connections between the column, reservoir and flow control adapter are spherical joints connected with CG-150 Stainless Steel pinchclamps.