Learn More
Chemglass Life Sciences Column, Chromatography, 2in ID X 12in E.L., 2mm Stpk
SDP
Catalog No. 501531038
Encompass_Preferred
Column, Chromatography, 2in ID X 12in E.L., 2mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.
Catalog No. 50-153-1038 Supplier Chemglass Life Sciences Supplier No. CG118719
Column, Chromatography, 2in ID X 12in E.L., 2mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.