Learn More
Cole-Parmer Cole-Parmer Column Clamp, Large, Stainless Steel, 2.52" to 5.51" Grip Range
Catalog No. NC0107232 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for large cylindrical glassware and similar labware. The large, flat thumbscrew/worm-drive provides a firm hold. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
Catalog No. NC0107232 Supplier Cole-Parmer Supplier No. 0806410
Ideal for large cylindrical glassware and similar labware. The large, flat thumbscrew/worm-drive provides a firm hold. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.