Learn More
Chemglass Life Sciences Clamp, Column, Large, 121mm to 140mm Clamping Range
SDP
Catalog No. CG922103
Encompass_Preferred
Clamp, Column, Large, 121mm to 140mm Clamping Range. Ideal for Chromatography columns and large cylindrical glassware, the column clamp is constructed of stainless steel. Large, flat thumb screw on the worm drive permits tightening and removing.
Catalog No. CG922103 Supplier Chemglass Life Sciences Supplier No. CG922103
Clamp, Column, Large, 121mm to 140mm Clamping Range. Ideal for Chromatography columns and large cylindrical glassware, the column clamp is constructed of stainless steel. Large, flat thumb screw on the worm drive permits tightening and removing.