Learn More
Ace Glass, Inc. Clamp, column, 92mm min to 165mm max, 368mm overall length with arm
SDP
Catalog No. 50980605
Encompass
Column clamp IDeal for use with cylindrical glassware having an outside diameter of 92mm to 165mm. A large, flat thumbscrew/worm-drive permits maximum tightening or removal in minimum time. The clamp is constructed of corrosion-resistant stainless steel, with an 11mm D x 211mm L rod extending from the clamp for easy mounting. 368mm overall length, including arm.
Catalog No. 50-980-605 Supplier Ace Glass, Inc. Supplier No. 1107808
Column clamp IDeal for use with cylindrical glassware having an outside diameter of 92mm to 165mm. A large, flat thumbscrew/worm-drive permits maximum tightening or removal in minimum time. The clamp is constructed of corrosion-resistant stainless steel, with an 11mm D x 211mm L rod extending from the clamp for easy mounting. 368mm overall length, including arm.