Learn More
Waters Corp Waters Corporation Column Support Clips, 10 per Pack
Catalog No. NC9700256 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Column support clips are essential spare parts designed to be used as spacers at fluidic connections to the column, ensuring even heat dissipation. These clips are supplied in convenient packs of 10.
  • Used as spacers at fluidic connections to the column.
  • Ensures even heat dissipation.
  • Supplied in a pack of 10.
Catalog No. NC9700256 Supplier Waters Corp Supplier No. 205000478
Column support clips are essential spare parts designed to be used as spacers at fluidic connections to the column, ensuring even heat dissipation. These clips are supplied in convenient packs of 10.
  • Used as spacers at fluidic connections to the column.
  • Ensures even heat dissipation.
  • Supplied in a pack of 10.