Learn More
Microsolv Technology Corporation Column Coupler .010 Universal
Catalog No. 5001451601
Encompass_Preferred
Union, Column Coupler, Universal Fit 0.25mm through Hole with 0.010"" ID tubing. Color Coded Blue. PEEK. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51601 Supplier Microsolv Technology Corporation Supplier No. 4745003
Union, Column Coupler, Universal Fit 0.25mm through Hole with 0.010"" ID tubing. Color Coded Blue. PEEK. MicroSolv Brand. 1/EA.