Learn More
Microsolv Technology Corporation Union, Column Coupler, Universal Fit 0.13mm through Hole with 0.005" ID tubing. Color Coded Red. PEEK. 1 EA.
Catalog No. 502271510
Encompass_Preferred
This convenient Column Connector is used to join two HPLC columns or a guard column and an analytical column in series. Sleeve is color coded Red to identify the tubing internal diameter and is made from polypropylene. Tubing is all PEEK material and is made from Super-T Tubing for precise concentricity and better performance.
This item is not returnable. View return policy
Catalog No. 50-227-1510 Supplier Microsolv Technology Corporation Supplier No. 4745001
This convenient Column Connector is used to join two HPLC columns or a guard column and an analytical column in series. Sleeve is color coded Red to identify the tubing internal diameter and is made from polypropylene. Tubing is all PEEK material and is made from Super-T Tubing for precise concentricity and better performance.