Learn More
Microsolv Technology Corporation Union, Column Coupler, Universal Fit 0.18mm through Hole with 0.007" ID tubing. Color Coded Yellow. PEEK. 1 EA.
Catalog No. 502271511
Encompass_Preferred
This convenient Column Connector is used to join two HPLC columns or a guard column and an analytical column in series. Sleeve is color coded Yellow to identify the tubing internal diameter and is made from polypropylene. Tubing is all PEEK material and is made from Super-T Tubing for precise concentricity and better performance.
This item is not returnable. View return policy
Catalog No. 50-227-1511 Supplier Microsolv Technology Corporation Supplier No. 4745002
This convenient Column Connector is used to join two HPLC columns or a guard column and an analytical column in series. Sleeve is color coded Yellow to identify the tubing internal diameter and is made from polypropylene. Tubing is all PEEK material and is made from Super-T Tubing for precise concentricity and better performance.