Learn More
Waters Corp Waters Column, C18, 1.7 µm, 130 Å, 2.1 x 100 mm
Catalog No. NC9051597 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CSH C18 columns provide alternate selectivity for UPLC applications, addressing peak shape asymmetry and poor loading capacity for basic compounds. Leveraging Charged Surface Hybrid (CSH) technology, they offer enhanced loading capacity, rapid re-equilibration after pH changes, and superior peak shape for basic compounds at low pH without needing TFA.
  • Provides alternate selectivity for reversed-phase UPLC applications.
  • Addresses peak shape asymmetry and poor loading capacity for basic compounds.
  • Features Charged Surface Hybrid (CSH) technology for enhanced loading capacity and rapid re-equilibration.
  • Offers superior peak shape for basic compounds, improved reproducibility, and excellent low pH stability.
  • Achieves TFA peak shape performance without TFA, utilizing MS-friendly mobile phase additives.
  • Manufactured in a cGMP, ISO 9001 certified facility with batch-specific testing.
Catalog No. NC9051597 Supplier Waters Corp Supplier No. 186005297
CSH C18 columns provide alternate selectivity for UPLC applications, addressing peak shape asymmetry and poor loading capacity for basic compounds. Leveraging Charged Surface Hybrid (CSH) technology, they offer enhanced loading capacity, rapid re-equilibration after pH changes, and superior peak shape for basic compounds at low pH without needing TFA.
  • Provides alternate selectivity for reversed-phase UPLC applications.
  • Addresses peak shape asymmetry and poor loading capacity for basic compounds.
  • Features Charged Surface Hybrid (CSH) technology for enhanced loading capacity and rapid re-equilibration.
  • Offers superior peak shape for basic compounds, improved reproducibility, and excellent low pH stability.
  • Achieves TFA peak shape performance without TFA, utilizing MS-friendly mobile phase additives.
  • Manufactured in a cGMP, ISO 9001 certified facility with batch-specific testing.