Learn More
Microsolv Technology Corporation Column Diamond Hydr 100X2.1Mm
Catalog No. 5001451319
Encompass_Preferred
HPLC Column, Diamond Hydride, 4um, 2.1mm ID x 100mm Length, 100A. Cogent TYPE-C Brand. 1/EA.
Catalog No. 50-014-51319 Supplier Microsolv Technology Corporation Supplier No. 7000010P2
HPLC Column, Diamond Hydride, 4um, 2.1mm ID x 100mm Length, 100A. Cogent TYPE-C Brand. 1/EA.