Learn More
Microsolv Technology Corporation Column Diamond Hydr 2.1X50Mm
Catalog No. 5001451315
Encompass_Preferred
HPLC Column, Diamond Hydride, 4um, 2.1mm ID x 50mm Length, 100A. Cogent TYPE-C Brand. 1/EA.
Catalog No. 50-014-51315 Supplier Microsolv Technology Corporation Supplier No. 7000005P2
HPLC Column, Diamond Hydride, 4um, 2.1mm ID x 50mm Length, 100A. Cogent TYPE-C Brand. 1/EA.