Learn More
Microsolv Technology Corporation Column End Plugs Blue
Catalog No. 5001451615
Encompass_Preferred
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Blue. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51615 Supplier Microsolv Technology Corporation Supplier No. 4902701B
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Blue. MicroSolv Brand. 10/PK.