Learn More
Microsolv Technology Corporation Column End Plugs Green
Catalog No. 5001451619
Encompass_Preferred
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Green. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51619 Supplier Microsolv Technology Corporation Supplier No. 4902705G
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Green. MicroSolv Brand. 10/PK.