Learn More
Microsolv Technology Corporation Column End Plugs Red
Catalog No. 5001451616
Encompass_Preferred
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Red. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51616 Supplier Microsolv Technology Corporation Supplier No. 4902702R
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Red. MicroSolv Brand. 10/PK.