Learn More
Microsolv Technology Corporation Column End Plugs White
Catalog No. 5001451618
Encompass_Preferred
End Plugs, for HPLC Columns. Nylon 10-32 Thread. White. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51618 Supplier Microsolv Technology Corporation Supplier No. 4902704W
End Plugs, for HPLC Columns. Nylon 10-32 Thread. White. MicroSolv Brand. 10/PK.