Learn More
Microsolv Technology Corporation Column End Plugs Yellow
Catalog No. 5001451617
Encompass_Preferred
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Yellow. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51617 Supplier Microsolv Technology Corporation Supplier No. 4902703Y
End Plugs, for HPLC Columns. Nylon 10-32 Thread. Yellow. MicroSolv Brand. 10/PK.