Learn More
Bio Rad Laboratories COLUMN FAST CARBOHYDRATE
Catalog No. 509012154
Encompass
Columns, Carbohydrate Analysis; Bio-Rad; Aminex; Fast carbohydrate; 100 x 7.8mm; Temperature: 85 deg.C; Flow rate: 0.6mL/min
Catalog No. 50-901-2154 Supplier Bio Rad Laboratories Supplier No. 1250105
Columns, Carbohydrate Analysis; Bio-Rad; Aminex; Fast carbohydrate; 100 x 7.8mm; Temperature: 85 deg.C; Flow rate: 0.6mL/min