Learn More
Trajan Scientific Column Fast QC1 PCB 1/EA
SDP
Catalog No. 5000494117
Encompass_Preferred
0 1 mm ID x 0 1 m film thickness x 10 m length fast HT8-PCB GC capillary column
Catalog No. 50-004-94117 Supplier Trajan Scientific Supplier No. 054690
0 1 mm ID x 0 1 m film thickness x 10 m length fast HT8-PCB GC capillary column