Learn More
Chemglass Life Sciences Glass Absorption Column ONLY, 12" OAL
SDP
Catalog No. 501531250
Encompass_Preferred
Glass Absorption Column ONLY, 12" OAL. Heavy wall column having 60mm Duran flanges on both ends. Column is 12 (300mm) long with an indent to hold the glass filled PTFE packing support near the bottom of the column. Packing support sold separately.
Catalog No. 50-153-1250 Supplier Chemglass Life Sciences Supplier No. CG183013
Glass Absorption Column ONLY, 12" OAL. Heavy wall column having 60mm Duran flanges on both ends. Column is 12 (300mm) long with an indent to hold the glass filled PTFE packing support near the bottom of the column. Packing support sold separately.