Learn More
Teledyne ISCO, Inc RediSep Gold C18 Reversed-Phase column, 150 gram.  Package of 1
Supplier:  Teledyne ISCO, Inc 692203338
Encompass_Preferred
Column, C18; Teledyne Isco; RediSep Rf Gold; High performance; Sample size: 130mg-3mg; Column size: 150g; 1 column/package
Catalog No. 50-200-825