Learn More
Teledyne ISCO, Inc COLUMN GOLD C18 415G
Catalog No. 50200827 Shop All Teledyne ISCO Inc. Products
Encompass_Preferred
Column, C18; Teledyne Isco; RediSep Rf Gold; High performance; Sample size: 360mg-8.3g; Column size: 415g; 1 column/package
Catalog No. 50-200-827 Supplier Teledyne ISCO, Inc Supplier No. 692203341
Column, C18; Teledyne Isco; RediSep Rf Gold; High performance; Sample size: 360mg-8.3g; Column size: 415g; 1 column/package