Learn More
Teledyne ISCO, Inc RediSep Rf Gold Normal Phase Silica Flash Columns. Column Type: Single use, disposable. 220 gram, package of 4.
Catalog No. 509018221 Shop All Teledyne ISCO Inc. Products
Encompass_Preferred
RediSep Rf Gold Normal-Phase Silica. High-performance spherical media for sharper peaks and faster purifications. Chemists choose our RediSep Rf Gold columns to take advantage of its patented silica media. Outstanding resolution to purify closely eluting compounds and isomers; faster purification runs to save time and solvent; reduced purification times to preserve silica-sensitive compounds; and more concentrated fractions to reduce post-run solvent evaporation time. Standard Features: High flow rates for fast purification of target compounds, Low back pressure, Reliable, consistent performance from automated precision packing, Column sizes available for purifying milligrams to several grams, RFID technology for automatic set up on compatible instruments, Fits all Isco CombiFlash automated organic purification instruments, Eliminates health hazard from inhaling silica gel dust, Long shelf life in airtight, resealable packages and Luer lock end fittings.
Catalog No. 50-901-8221 Supplier Teledyne ISCO, Inc Supplier No. 692203359
RediSep Rf Gold Normal-Phase Silica. High-performance spherical media for sharper peaks and faster purifications. Chemists choose our RediSep Rf Gold columns to take advantage of its patented silica media. Outstanding resolution to purify closely eluting compounds and isomers; faster purification runs to save time and solvent; reduced purification times to preserve silica-sensitive compounds; and more concentrated fractions to reduce post-run solvent evaporation time. Standard Features: High flow rates for fast purification of target compounds, Low back pressure, Reliable, consistent performance from automated precision packing, Column sizes available for purifying milligrams to several grams, RFID technology for automatic set up on compatible instruments, Fits all Isco CombiFlash automated organic purification instruments, Eliminates health hazard from inhaling silica gel dust, Long shelf life in airtight, resealable packages and Luer lock end fittings.