Learn More
Waters Corp Waters Column Heater Module, Four Column Capacity, 30 cm Column Length
Catalog No. NC2868614 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Column Heater Module is designed to house up to four columns (up to 30 centimeters in length) or one column with a Ninhydrin reaction coil. It supports simultaneous and independent control by the Temperature Control Module II. Measured temperatures, set points, and limits are programmable via the front panel or through Empower™ or Millennium®32 software.
  • Houses up to four columns (up to 30 cm) or one column with a Ninhydrin reaction coil.
  • Supports up to three columns and one RXN 1000 reaction coil.
  • Offers independent control via Temperature Control Module II.
  • Displays measured temperatures on the front panel.
  • Allows programming of temperature set points and limits.
  • Provides quick access to settings with a single keystroke.
  • Compatible with Empower™ or Millennium®32 software control.
Catalog No. NC2868614 Supplier Waters Corp Supplier No. WAT038040
The Column Heater Module is designed to house up to four columns (up to 30 centimeters in length) or one column with a Ninhydrin reaction coil. It supports simultaneous and independent control by the Temperature Control Module II. Measured temperatures, set points, and limits are programmable via the front panel or through Empower™ or Millennium®32 software.
  • Houses up to four columns (up to 30 cm) or one column with a Ninhydrin reaction coil.
  • Supports up to three columns and one RXN 1000 reaction coil.
  • Offers independent control via Temperature Control Module II.
  • Displays measured temperatures on the front panel.
  • Allows programming of temperature set points and limits.
  • Provides quick access to settings with a single keystroke.
  • Compatible with Empower™ or Millennium®32 software control.