Learn More
Waters Corp Waters Column Heater Inserts, 4 mm I.D., 30 cm Length
Catalog No. 50793496 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These column heater inserts are designed to provide consistent temperature control for chromatographic separations. They enable the use of specific column dimensions within compatible heating systems.
  • Designed for 4 mm I.D. columns.
  • Accommodates columns up to 30 cm in length.
  • Ensures precise temperature management in column heater modules.
Catalog No. 50-793-496 Supplier Waters Corp Supplier No. WAT019887
These column heater inserts are designed to provide consistent temperature control for chromatographic separations. They enable the use of specific column dimensions within compatible heating systems.
  • Designed for 4 mm I.D. columns.
  • Accommodates columns up to 30 cm in length.
  • Ensures precise temperature management in column heater modules.