Learn More
Advanced Materials Technology HALO 400 Å ES-C18, 3.4 µm, 2.1 x 50 mm
Supplier:  Advanced Materials Technology 93412402
Encompass_Preferred
HALO BIOCLASS ES-C18 400 Å Protein columns made with innovative large pore Fused-Core particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO 400 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Catalog No. 50-105-0909