Learn More
Trajan Scientific Column HT5 QC2 Medical 1/EA
SDP
Catalog No. 5000494101
Encompass_Preferred
0 22 mm ID x 0 1 m film thickness x 25 m length HT5 GC capillary column
Catalog No. 50-004-94101 Supplier Trajan Scientific Supplier No. 054632
0 22 mm ID x 0 1 m film thickness x 25 m length HT5 GC capillary column